Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant NAMPT (from <i>E. coli</i>)

Human recombinant NAMPT (from E. coli)

50 µG
Catalog # PRSI90-010
Supplier:   ProSci Inc.
Human recombinant NAMPT (from <i>E. coli</i>)
Catalog # PRSI90-010
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant NAMPT (from E. coli)

50 µG

CAS Number:  
Price
599,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Nicotinamide phosphoribosyltransferase (Nampt; pre-B cell colony-enhancing factor; PBEF; Visfatin) is an 52kDa adipokine secreted by adipose tissue and involved in the biosynthesis of nicotinamide adenine dinucleotide (NAD+). Two forms of Nampt exist, an intracellular form (iNampt) and an extracellular form (eNampt). While the function of iNampt as an essential and rate-limiting NAD+ biosynthetic enzyme is well established, the physiological role of eNampt is still a matter of debate. Nampt has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    PRSI90-010
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    NAMPT
  • Storage conditions:
    Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Formulation:
    Lyophilized, Salt-free.
  • Protein synonyms:
    Pre-B Cell Colony Enhancing Factor 1|Nicotinamide Phosphoribosyltransferase|PBEF1
  • Purity:
    >90% (SDS-PAGE)
  • Tag sequence:
    His Tag
  • Sequence:
    1-491
  • Species:
    Human
  • Source:
    E. coli
  • Biological activity:
    Shows adipogenic effects in stimulated differentiating 3T3-L1 cells.

Specifications