Order Entry
Export
ContactUsLinkComponent
Mouse recombinant Fetuin A
Mouse recombinant Fetuin A
Catalog # PRSI92-714
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant Fetuin A
Catalog # PRSI92-714
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
  • Species:
    Mouse
  • Environmentally Preferable:
  • Tag sequence:
    C-10 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ahsg|Fetua|Alpha-2-HS-glycoprotein|Countertrypin|Fetuin-A
  • Protein/peptide name:
    Fetuin A
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    36.7 kD
  • Sequence:
    Ala19-Ile345
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris, 150mM NaCl, pH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Alpha-2-HS-glycoprotein (AHSG) is a glycoprotein that is composed of two subunits, the A and B chains, belongs to the Cystatin family of proteases inhibitors. It is highly expressed in embryonic cells and adult hepatocytes, and is expressed to a lesser extent in monocytes/macrophages. AHSG is an important circulating inhibitor of calcification in vivo, and is downregulated during the acute-phase response. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. In addition, AHSG may influence the resolution of inflammation by modulating the phagocytosis of apoptotic cells by macrophages. ASHG blocks TGF-beta-dependent signalling in osteoblastic cells.

Fusion-Tag: C-10 His tag

This recombinant protein can be used for biological assays. For research use only.