Order Entry
Export
ContactUsLinkComponent
Human recombinant CHRNB3 (from cells)
Human recombinant CHRNB3 (from cells)
Catalog # PRSI92-338
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CHRNB3 (from cells)
Catalog # PRSI92-338
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Neuronal acetylcholine receptor subunit beta-3
  • Protein/peptide name:
    CHRNB3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.3 kD
  • Sequence:
    Ile25-Leu232
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Neuronal acetylcholine receptor subunit beta-3(CHRNB3) is a cell membrane protein and belongs to the ligand-gated ion channel (TC 1.A.9) family. CHRNB3 seems to be composed of two different type of subunits: alpha and beta. The CHRNB3 are (hetero) pentamers composed of homologous subunits. The subunits that make up the muscle and neuronal forms of CHRNB3 are encoded by separate genes and have different primary structure. There are several subtypes of neuronal CHRNB3 that vary based on which homologous subunits are arranged around the central channel. They are classified as alpha-subunits if like muscle alpha-1, they have a pair of adjacent cysteines as part of the presumed acetylcholine binding site. Subunits lacking these cysteine residues are classified as beta-subunits.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.