Mouse recombinant L-selectin
PRSI92-322
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:LECAM1|Lymph node homing receptor|Sell|Leukocyte adhesion molecule 1|CD62 antigen-like family member L|CD62L|Lymphocyte antigen 22|L-selectin
- Protein/peptide name:L-selectin
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:34.1 kD
- Sequence:Trp39-Asn332
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
L-Selectin is a member of a family of Selectin that is transiently expressed on vascular endothelial cells in response to IL-1 beta and TNF-alpha. L-Selectin (Leukocyte Selectin, LAM-1, LECAM-1, LECCAM-1, TQ1, Leu-8, MEL-14 antigen, DREG, lymph node homing receptor, CD62L) is expressed constitutively on a wide variety of leukocytes and mediates a number of leukocyte-endothelial interactions, including the binding of lymphocytes to HEV of peripheral lymph node high endothelial venules (HEV), neutrophil rollling, and leukocyte attachment to cytokine-treated endothelium in vitro.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.