Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant NCR3

Human recombinant NCR3

50 µG
Catalog # PRSI92-151
Supplier:   ProSci Inc.
Human recombinant NCR3
Catalog # PRSI92-151
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant NCR3

50 µG

CAS Number:  
Price
730,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Natural Cytotoxicity Triggering Receptor 3 (NCR3) along with NKp44 and NKp46 constitute a group of receptors termed “Natural Cytotoxicity Receptors”. They play a major role in triggering NK-mediated killing of most tumour cells lines. NKp30 is a type I transmembrane protein having a single extracellular V-like immunoglobulin domain. NKp30 is selectively expressed both in resting and activated human NK cells. In addition, NKp30 is also involved in NK-mediated induction of dendritic cell (DC) maturation. It has been demonstrated that NK cell activation signalling specifically induces lytic activity against several tumour cell types and synthesis of new NF-κB dependent proteins during the initiation of cytotoxicity.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    PRSI92-151
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    NCR3
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Protein synonyms:
    CD337|1C7|Natural Cytotoxicity Triggering Receptor 3|LY117|NCR3|Natural Killer Cell p30-Related Protein|NK-p30|Activating Natural Killer Receptor p30|NKp30
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Tag sequence:
    C-Fc tag
  • Sequence:
    Leu19-Thr138
  • Tested applications:
    Bioassay
  • Species:
    Human
  • Molecular weight:
    40.2 kD

Specifications