Order Entry
Export
ContactUsLinkComponent
Human recombinant CLIC1 (from E. coli)
Human recombinant CLIC1 (from E. coli)
Catalog # PRSI92-150
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CLIC1 (from E. coli)
Catalog # PRSI92-150
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    G6|hRNCC|Nuclear Chloride Ion Channel 27|CLIC1|NCC27|Regulatory Nuclear Chloride Ion Channel Protein|Chloride Channel ABP|Chloride Intracellular Channel Protein 1
  • Protein/peptide name:
    CLIC1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    29 kD
  • Sequence:
    Met1-Lys241
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Chloride Intracellular Channel Protein 1 (CLIC1) belongs to the Chloride Channel CLIC family and contains one GST C-terminal domain. CLIC1 can be expressed in various cell types, but it is especially prominent in the heart, placenta, liver, kidney, and pancreas. CLIC1 can insert into membranes and form chloride ion channels. The channel activity depends on the pH. CLIC1 membrane insertion seems to be redox-regulated and may occur only under oxidising conditions. CLIC1 is also involved in the regulation of the cell cycle.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.