Human recombinant PDCD5 (from E. coli)
PRSI92-144
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:PDCD5|TFAR19|Programmed Cell Death Protein 5|TF-1 Cell Apoptosis-Related Protein 19|Protein TFAR19
- Protein/peptide name:PDCD5
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:16.4 kD
- Sequence:Met1-Tyr125
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Programmed Cell Death Protein 5 (PDCD5) is a member of the PDCD5 family. PDCD5 is expressed in tumour cells during apoptosis, independent of apoptosis-inducing stimuli. This protein may function in the process of apoptosis. PDCD5 is upregulated during apoptosis where it translocates rapidly from the cytoplasm to the nucleus. PDCD5 may play an important regulator of K (lysine) acetyltransferase 5 (a protein involved in transcription, DNA damage response and cell cycle control) by inhibiting its proteasome-dependent degradation. PDCD5 is an important novel protein that regulates both apoptotic and non-apoptotic programmed cell death.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.