Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant TMIGD2 (from cells)

Human recombinant TMIGD2 (from cells)

50 µG
Catalog # PRSI91-907
Supplier:   ProSci Inc.
Human recombinant TMIGD2 (from cells)
Catalog # PRSI91-907
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant TMIGD2 (from cells)

50 µG

CAS Number:  
Price
730,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

TMIGD2 is a single-pass type I membrane protein, which contains one Ig-like (immunoglobulin-like) domain. It is widely expressed in many tissues, such as epithelial, endothelial cells and lung. However, it isn’t detected in thyroid, cerebellum, thymus and cerebral cortex. TMIGD2 can form homophilic interactions that could regulate cell-cell interaction. It Interacts with CACNB2, DST, MIA and NCKIPSD. It is shown that TMIGD2 plays a role in cell-cell interaction, cell migration, and angiogenesis.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Protein/peptide name:
    TMIGD2
  • Protein synonyms:
    Transmembrane and immunoglobulin domain-containing protein 2|Immunoglobulin and proline-rich receptor 1|TMIGD2|IGPR1|CD28H|IGPR-1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Leu23-Gly150
  • Source:
    Cells
  • Molecular weight:
    17.3 kD
  • Cat. No.:
    PRSI91-907
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Tested applications:
    used for biological assays
  • Species:
    Human

Specifications