Human recombinant LMIR2 (from cells)
PRSI91-890
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:CMRF35|Immunoglobulin Superfamily Member 16|CLM-6|CMRF35-A1|CD300 Antigen-like Family Member C|IgSF16|CMRF-35|CMRF35-like Molecule 6|CD300c|CMRF35A|CMRF35A1
- Protein/peptide name:LMIR2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:44.9 kD
- Sequence:Gly21-Arg183
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
CD300C is a single-pass type I membrane protein which belongs to the immunoregulatory signaling (IRS) family. CD300C contains one Ig-like V-type domain and is present on the surface of natural killer cells, granulocytes, most myeloid cells, dendritic cells, and a subpopulation of T and B lymphocytes. The CD300C (CMRF-35A) and CD300A (CMRF-35H) molecules are homologous leukocyte surface proteins. CD300a and CD300C play an important role in the cross-regulation of TNF-alpha and IFN-alpha secretion from pDCs. CD300A and CD300C are indistinguishable on the surface of NK cells. The ligand for CD300C is presently unknown.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag