Order Entry
Export
ContactUsLinkComponent
Human recombinant CD99 antigen-like 2 (from cells)
Human recombinant CD99 antigen-like 2 (from cells)
  PRSI91-768
 :  
Human recombinant CD99 antigen-like 2 (from cells)
  PRSI91-768
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CD99|MIC2-like Protein 1|CD99L2|CD99 antigen-like Protein 2|MIC2L1
  • Protein/peptide name:
    CD99 antigen-like 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.4 kD
  • Sequence:
    Asp26-Ala188
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

CD99 Antigen-Like Protein 2 (CD99L2) belongs to the CD99 family. CD99L2 is a single-pass type I membrane protein and expressed in many tissues, with low expression in thymus. CD99L2 plays a role in a late step of leukocyte extravasation helping cells to overcome the endothelial basement membrane. CD99L2 and CD99 are involved in trans-endothelial migration of neutrophils in vitro and in the recruitment of neutrophils into inflamed peritoneum. A similar protein in mouse functions as an adhesion molecule during leukocyte extravasation. Alternate splicing results in multiple transcript variants.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag