Human recombinant INSL4 (from cells)
Catalog # PRSI91-712
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:EPIL|Insulin-like Peptide 4|INSL4|Early Placenta Insulin-like Peptide|Placentin
- Protein/peptide name:INSL4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:13.6 kD
- Sequence:Ala26-Thr139
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NACl,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Early Placenta Insulin-Like Peptide (INSL4) belongs to the insulin family. INSL4 is expressed in the early placental cytotrophoblast and syncytiotrophoblast INSL4 is a secreted protein and a precursor that undergoes post-translational cleavage to produce 3 polypeptide chains, A-C, that form tertiary structures composed of either all three chains, or just the A and B chains. INSL4 plays an important role in the development of trophoblast and in the regulation of bone formation.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag