Human recombinant protamine-2 (from cells)
50 µG
Catalog # PRSI91-653
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
Ribonucleoside-Diphosphate Reductase Subunit M2 (RRM2) belongs to the ribonucleoside diphosphate reductase small chain family. The reductase of RRM2 catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the encoded protein (M2) is regulated in a cell-cycle dependent fashion. RRM2 supplies the precursors essential for DNA synthesis. RRM2 catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. Phosphorylation on Ser-20 relieves the inhibitory effect on Wnt signaling.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Documents
75789-966.pdf
Datasheet
Specifications
Specifications
- Pk:50 µG
- Protein/peptide name:Protamine-2
- Protein synonyms:Ribonucleoside-Diphosphate Reductase Subunit M2|RR2|RRM2|Ribonucleotide Reductase Small Chain|Ribonucleotide Reductase Small Subunit|PRM2
- Purity:> 95% as determined by reducing SDS-PAGE
- Sequence:Met1-Phe389
- Source:Cells
- Molecular weight:45.9 kD
- Cat. No.:PRSI91-653
- Protein/peptide type:Recombinant
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tag sequence:C-6 His tag
- Tested applications:Bioassay
- Species:Human



