Order Entry
Export
ContactUsLinkComponent
Human recombinant serpin A3 (from cells)
Human recombinant serpin A3 (from cells)
Catalog # PRSI91-484
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant serpin A3 (from cells)
Catalog # PRSI91-484
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Cell Growth-Inhibiting Gene 24/25 Protein|Serpin A3|AACT|Alpha-1-Antichymotrypsin|ACT|SERPINA3
  • Protein/peptide name:
    Serpin A3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    46.33 kD
  • Sequence:
    His24-Ala423
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM HEPES, 150mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Serpin A3 belongs to the Serpin superfamily of serine protease inhibitors. Serpin A3 has been shown to inhibit some serine proteases, such as neutrophil cathepsin G and mast cell chymase. Serpin A3 is synthesized initially in the liver and secreted in plasma. Serpin A3 has been found in the amyloid plaques from the hippocampus of Alzheimer disease brains. In addition to, Serpin A3 is associated with liver disease and Parkinson disease and chronic obstructive pulmonary disease.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag