Order Entry
Export
ContactUsLinkComponent
Human recombinant Chymotrypsinogen (from cells)
Human recombinant Chymotrypsinogen (from cells)
Catalog # PRSI91-443
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Chymotrypsinogen (from cells)
Catalog # PRSI91-443
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    CTRB|CTRB1|Chymotrypsinogen B
  • Protein/peptide name:
    Chymotrypsinogen
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26.89 kD
  • Sequence:
    Cys19-Asn263
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Chymotrypsinogen B (CTRB1) is a 263 amino acid protein with signal peptide (1-18) and Chymotrypsinogen B (19-263). Chymotrypsinogen B have three chains:Chymotrypsin B chain A(19-31), Chymotrypsin B chain B(34-164), Chymotrypsin B chain C (167-263). Chymotrypsinogen B is a Serine Protease Hydrolase secrets into gastrointestinal tract as the inactive precursor Chymotrypsinogen.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag