Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant HGPRT (from <i>E. coli</i>)

Human recombinant HGPRT (from E. coli)

50 µG
Catalog # PRSI91-274
Supplier:   ProSci Inc.
Human recombinant HGPRT (from <i>E. coli</i>)
Catalog # PRSI91-274
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant HGPRT (from E. coli)

50 µG

CAS Number:  
Price
757,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Hypoxanthine-Guanine Phosphoribosyltransferase (HGPRT) has an important role in the generation of purine nucleotides through the purine salvage pathway. HPRT1 functions to salvage purines from degraded DNA to renewed purine synthesis, it acts as a catalyst in the reaction between guanine and phosphoribosyl pyrophosphate to form GMP.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Protein/peptide name:
    HGPRT
  • Protein synonyms:
    HGPRTase|HGPRT|HPRT1|HPRT|Hypoxanthine-Guanine Phosphoribosyltransferase
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Ala218
  • Source:
    E. coli
  • Molecular weight:
    27.79 kD
  • Cat. No.:
    PRSI91-274
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris, 250mM NaCl, 2mM EDTA, 30% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N-6 His tag
  • Tested applications:
    For Biological Assays
  • Species:
    Human

Specifications