Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant HSF2 (from <i>E. coli</i>)

Human recombinant HSF2 (from E. coli)

50 µG
Catalog # PRSI91-264
Supplier:   ProSci Inc.
Human recombinant HSF2 (from <i>E. coli</i>)
Catalog # PRSI91-264
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant HSF2 (from E. coli)

50 µG

CAS Number:  
Price
730,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Heat Shock Factor Protein 2 (HSF2) belongs to the HSF family of transcription factors that bind specifically to the heat-shock promoter element and activate transcription. In higher eukaryotes, HSF is unable to bind to the HSE unless the cells are heat shocked. HSF2 is widely expressed in many cells and tissues. HSF2 is located on Cytoplasmic during normal growth. But when it is activited, HSF2 moves to the nucleus.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Protein/peptide name:
    HSF2
  • Protein synonyms:
    HSF2|HSTF2|Heat Shock Transcription Factor 2|Heat Shock Factor Protein 2|HSTF 2|HSF 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Ser411-Ser536
  • Source:
    E. coli
  • Molecular weight:
    15.9 kD
  • Cat. No.:
    PRSI91-264
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human

Specifications