Order Entry
Export
Orders LinkContactUsLinkComponent
 

Human recombinant AIF (from E. coli)

50 µG
  PRSI91-252
Human recombinant AIF (from <i>E. coli</i>)
  PRSI91-252
 :  
Human recombinant AIF (from E. coli)

50 µG

 :  
 
730,00 €
   
1 * 50 µG
 

Apoptosis-Inducing Factor 1, Mitochondrial (AIFM1) is a flavoprotein essential for nuclear disassembly in apoptotic cells that is found in the mitochondrial intermembrane space in healthy cells. During apoptosis, it is translocated from the mitochondria to the nucleus to function as a proapoptotic factor in a caspase-independent pathway, while in normal mitochondria, it functions as an antiapoptotic factor via its oxidoreductase activity. The soluble form (AIFsol) found in the nucleus induces parthanatos i.e., caspase-independent fragmentation of chromosomal DNA. AIFM1 interacts with EIF3G, and thereby inhibits the EIF3 machinery and protein synthesis, and activates casapse-7 to amplify apoptosis. It binds to DNA in a sequence-independent manner and plays a critical role in caspase-independent, pyknotic cell death in hydrogen peroxide-exposed cells.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

 

 

  • Pk:
    50 µG
  • Protein/peptide name:
    AIF
  • Protein synonyms:
    AIF|Apoptosis-Inducing Factor 1 Mitochondrial|AIFM1|Programmed Cell Death Protein 8|PDCD8
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Glu121-Asp613
  • Source:
    E. coli
  • Molecular weight:
    56.2 kD
  • Cat. No.:
    PRSI91-252
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human