Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant UROIIIS (from <i>E. coli</i>)

Human recombinant UROIIIS (from E. coli)

50 µG
Catalog # PRSI91-248
Supplier:   ProSci Inc.
Human recombinant UROIIIS (from <i>E. coli</i>)
Catalog # PRSI91-248
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant UROIIIS (from E. coli)

50 µG

CAS Number:  
Price
757,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Uroporphyrinogen-III Synthase is an enzyme which belongs to the uroporphyrinogen-III synthase family. Uroporphyrinogen-III Synthase is ubiquitous and it is involved in Porphyrin metabolism. Porphyrins act as cofactors for a multitude of enzymes that perform a variety of processes within the cell such as Methionine synthesis (Vitamin B12) or oxygen transport (Heme). Uroporphyrinogen-III Synthase can catalyze cyclization of the linear Tetrapyrrole, Hydroxymethylbilane, to the Macrocyclic Uroporphyrinogen III, the branch point for the various sub-pathways leading to the wide diversity of Porphyrins. Defects in Uroporphyrinogen-III Synthase are the cause of Congenital Erythropoietic Porphyria (CEP).

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Protein/peptide name:
    UROIIIS
  • Protein synonyms:
    UROIIIS|Hydroxymethylbilane Hydrolyase [Cyclizing]|UROS|Uroporphyrinogen-III Cosynthase|Uroporphyrinogen-III Synthase
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Pro277
  • Source:
    E. coli
  • Molecular weight:
    29.7 kD
  • Cat. No.:
    PRSI91-248
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human

Specifications