Order Entry
Export
ContactUsLinkComponent
Human recombinant ZBTB17 (from E. coli)
Human recombinant ZBTB17 (from E. coli)
Catalog # PRSI91-217
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ZBTB17 (from E. coli)
Catalog # PRSI91-217
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    ZBTB17|Zinc Finger Protein 60|MIZ1|Miz-1|Zinc Finger Protein 151|ZNF151|Zinc Finger and BTB Domain-Containing Protein 17|Myc-Interacting Zinc Finger Protein 1|ZNF60
  • Protein/peptide name:
    ZBTB17
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    22.33 kD
  • Sequence:
    Met1-Ala188
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.25. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Zinc Finger and BTB Domain-Containing Protein 17 (ZBTB17) belongs to the Kruppel C2H2-type zinc finger protein family. ZBTB17 may function as a housekeeping DNA-binding protein that regulates the expression of specific genes, it has been shown to bind to the promoters of adenovirus major late protein and cyclin D1 and activate transcription. ZBTB17 may has growth arrest activity, probably through inhibition of cell cycle progression. ZBTB17 required for early embryonic development during gastrulation. ZBTB17 induces cell arrest at G1, an effect mediated by its activation of the gene coding for P15INK4b. This effect is blocked by Myc, which displaces transcriptional coactivators bound to ZBTB17. Although the downregulation of ZBTB17 may contribute to Myc-induced cell transformation, the de-activation of ZBTB17 is absolutely essential for Myc-induced apoptosis.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag