Human Recombinant CCL26 (from E. coli)
Product Details & Documents
Chemokine Ligand 26 protein (CCL26) is a novel small cytokine belonging to the CC chemokine family which is involved in immunoregulatory and inflammatory processes. CCL26 is constitutively expressed in thymus, but only transiently expressed in phytohemagglutinin-stimulated peripheral blood mononuclear cells. It specifically binds and induces chemotaxis in T cells and elicits its effects by interacting with the chemokine receptor CCR4. CCL26, along with Eotaxin-1 and Eotaxin-2, selectively activates the CC chemokine receptor 3 (CCR3). The Eotaxin-3-CCR3 interaction may play an important role in allergic diseases such as atopic dermatitis and bronchial asthma. The full-length cDNA for CCL26 encodes a protein of 94 amino acids with a putative signal peptide of either 23 or 26 amino acid residues. Both the 71 and 68 amino acid residue variants of recombinant CCL26 demonstrate equal potency in inducing chemotaxis of a human CCR3-transfected cell line. Unlike most other CC chemokines, CCL26 maps to human chromosome 7q11.2, within 40 kilobases of the Eotaxin-2 loci. CCL26 and Eotaxin-2 are unique in that they are the only chemokines identified to date that map to chromosome 7.
- This recombinant protein can be used for biological assays. For research use only.
Documents
75788-916.pdf
Datasheet
Specifications
Specifications
- Pk:50 µG
- Cat. No.:PRSI91-096
- Protein/peptide type:Recombinant
- Protein/peptide name:CCL26
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris, 1mM EDTA, 20% Glycerol, pH 9.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Protein synonyms:MIP-4-Alpha|Macrophage Inflammatory Protein 4-Alpha|TSC-1|Small-Inducible Cytokine A26|Thymic Stroma Chemokine-1|C-C Motif Chemokine 26|Eotaxin-3|SCYA26|CC Chemokine IMAC|CCL26
- Purity:> 95% as determined by reducing SDS-PAGE
- Sequence:Ala2-Gln290
- Tested applications:For Biological assays
- Species:Human
- Source:E. coli
- Molecular weight:8.53 kD



