Order Entry
Export
ContactUsLinkComponent
Human Recombinant CCL3 (from E. coli)
Human Recombinant CCL3 (from E. coli)
Catalog # PRSI91-048
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant CCL3 (from E. coli)
Catalog # PRSI91-048
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biological activity:
    ED50 is 3-10 ng/mL as determined by the ability of Recombinant CCL3 to chemoattract human CCR5 transfected BaF3 mouse proB cells.
  • Protein synonyms:
    MIP-1-Alpha|CCL3|G0S19-1|SCYA3|PAT 464.1|SIS-Beta|MIP1A|Small-Inducible Cytokine A3|C-C Motif Chemokine 3|G0/G1 Switch Regulatory Protein 19-1|Macrophage Inflammatory Protein 1-Alpha|MIP-1 alpha|Tonsillar Lymphocyte LD78 Alpha Protein
  • Protein/peptide name:
    CCL3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    7.5 kD
  • Sequence:
    Ser24-Ala92
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Human Chemokine (C-C Motif) Ligand 3 (CCL3) is a small cytokine belonging to the CC chemokine family. CCL3 is primarily expressed in T cells, B cells, and monocytes after antigen or mitogen stimulation. CCL3 exhibits chemoattractive and adhesive effects on lymphocytes. CCL3 exerts multiple effects on hematopoietic precursor cells and inhibits the proliferation of hematopoietic stem cells in vitro as well as in vivo. CCR1 and CCR5 have been identified as functional receptors for CCL3.

  • This recombinant protein can be used for biological assays. For research use only.