Order Entry
Export
ContactUsLinkComponent
Human recombinant PDL19 (from HEK293 cells)
Human recombinant PDL19 (from HEK293 cells)
Catalog # PRSI96-599
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant PDL19 (from HEK293 cells)
Catalog # PRSI96-599
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA.
    Immobilized Human PD-L1, Strep Tag at 5 ug/mL ( 100 ul/well ) can bind Biotinylated Human PD-1, His Tag & Fc tag with a linear range of 2-60 ng/mL.
  • Protein synonyms:
    B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
  • Protein/peptide name:
    PDL19
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    28.2 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

Programmed cell death 1 ligand 1 (PD-L1) is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses. B7-H1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. PD-L1 is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 involve in the costimulatory signal, essential for T-cell proliferation and production of IL10 and IFNG, in an IL2-dependent and a PDCD1-independent manner.