Order Entry
Export
ContactUsLinkComponent
Human recombinant FGF10 (from E. coli)
Human recombinant FGF10 (from E. coli)
Catalog # PRSI40-587
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant FGF10 (from E. coli)
Catalog # PRSI40-587
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    Determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors. The expected ED50 is = 0.5 ng/mL corresponding to a specific activity of = 2 x 106 units/mg.
  • Protein synonyms:
    Fibroblast growth factor 10|FGF-10|Keratinocyte growth factor 2
  • Protein/peptide name:
    FGF10
  • Purity:
    >95%
  • Formulation:
    Lyophilized

Specifications

About this item

FGF-10 is a heparin binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-10 is most related to KGF/FGF-7 and is expressed during the development and preferentially in adult lungs. It signals through the FGFR 2b. Recombinant human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues.

  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.