Human recombinant Atp6V1E1 (from E. coli)
Catalog # PRSIXW-RP3020
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:ATP6V1E|Vma4|ATP6E2|ATP6E|P31|V-type proton ATPase subunit E 1|V-ATPase 31 kDa subunit|V-ATPase subunit E 1
- Protein/peptide name:Atp6V1E1
- Purity:>95%
- Molecular weight:26 kDa (Calculated)
- Tested applications:ELISA, Western Blot, MS
Specifications
About this item
This protein is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, and receptor-mediated endocytosis. V-ATPase is comprised of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of a hexamer of three A and three B subunits plus the C, D, and E subunits. It contains the ATP catalytic site. The protein is known as the E subunit and is found ubiquitously.
- This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.