Human SDHAF1 peptide (18 aa near C-terminus)
PRSI6879P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near C-terminus
- Protein/peptide name:SDHAF1 Peptide (18 aa near C-terminus)
- Tested applications:Blocking
SDHAF1 peptide is used for blocking the activity of SDHAF1 antibody.