Human LMX1A peptide (16 aa near C-terminus)
PRSI7087P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near C-terminus
- Protein/peptide name:LMX1A Peptide (16 aa near C-terminus)
- Tested applications:Blocking
LMX1A Peptide is used for blocking the activity of LMX1A antibody.