Order Entry
Export
ContactUsLinkComponent
Human ESRRB peptide (19 aa near N-terminus)
Human ESRRB peptide (19 aa near N-terminus)
  PRSI6813P
 :  
undefined
Human ESRRB peptide (19 aa near N-terminus)
  PRSI6813P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    19 aa near N-terminus
  • Protein/peptide name:
    ESRRB Peptide (19 aa near N-terminus)
  • Tested applications:
    Blocking

 

 

ESRRB peptide is used for blocking the activity of ESRRB antibody.