Human PIERCE1 peptide (16 aa near N-terminus)
PRSI6721P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near N-terminus
- Protein/peptide name:PIERCE1 Peptide (16 aa near N-terminus)
- Tested applications:Blocking
PIERCE1 peptide is used for blocking the activity of PIERCE1 antibody.