Human Strumpellin peptide (19 aa near C-terminus)
PRSI6483P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near C-terminus
- Protein/peptide name:Strumpellin Peptide (19 aa near C-terminus)
- Tested applications:Blocking
DDA peptide is used for blocking the activity of DDA antibody.