Order Entry
Export
ContactUsLinkComponent
Human ZIP10 peptide (18 aa in the center)
Human ZIP10 peptide (18 aa in the center)
  PRSI6099P
 :  
Human ZIP10 peptide (18 aa in the center)
  PRSI6099P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 aa in the center
  • Protein/peptide name:
    ZIP10 Peptide (18 aa in the center)
  • Tested applications:
    Blocking

 

 

ZIP10 peptide is used for blocking the activity of ZIP10 antibody.