Human ZEB2 peptide (18 aa near C-terminus)
Catalog # PRSI5827P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near C-terminus
- Protein/peptide name:ZEB2 Peptide (18 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
ZEB2 peptide is used for blocking the activity of ZEB2 antibody.