Human SIRT1 peptide (19 aa near N-terminus)
PRSI5765P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near N-terminus
- Protein/peptide name:SIRT1 Peptide (19 aa near N-terminus)
- Tested applications:Blocking
SIRT1 peptide is used for blocking the activity of SIRT1 antibody.