Human APC11 peptide (18 aa in the center)
Catalog # PRSI5737P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa in the center
- Protein/peptide name:APC11 Peptide (18 aa in the center)
- Tested applications:Blocking
Specifications
About this item
APC11 peptide is used for blocking the activity of APC11 antibody.