Human APC10 peptide (16 aa in the center)
PRSI5735P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa in the center
- Protein/peptide name:APC10 Peptide (16 aa in the center)
- Tested applications:Blocking
APC10 peptide is used for blocking the activity of APC10 antibody.