Order Entry
Export
ContactUsLinkComponent
 
Human CCDC69A peptide (16 aa near N-terminus)
  PRSI5691P
 :  
undefined
Human CCDC69A peptide (16 aa near N-terminus)
  PRSI5691P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    16 aa near N-terminus
  • Protein/peptide name:
    CCDC69A Peptide (16 aa near N-terminus)
  • Tested applications:
    Blocking

 

 

CCDC69 peptide is used for blocking the activity of CCDC69 antibody.