Human ZC3H12A peptide (17 aa in the center)
PRSI5673P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa in the center
- Protein/peptide name:ZC3H12A Peptide (17 aa in the center)
- Tested applications:Blocking
ZC3H12A peptide is used for blocking the activity of ZC3H12A antibody.