Human TMEM16A peptide (17 aa near N-terminus)
PRSI5419P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa near N-terminus
- Protein/peptide name:TMEM16A Peptide (17 aa near N-terminus)
- Tested applications:Blocking
TMEM16A peptide is used for blocking the activity of TMEM16A antibody.