Human AGTR1 peptide (16 aa in the center)
PRSI5391P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa in the center
- Protein/peptide name:AGTR1 Peptide (16 aa in the center)
- Tested applications:Blocking
AGTR1 peptide is used for blocking the activity of AGTR1 antibody.