Human CIP75 peptide (18 aa near N-terminus)
Catalog # PRSI5267P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near N-terminus
- Protein/peptide name:CIP75 Peptide (18 aa near N-terminus)
- Tested applications:Blocking
Specifications
About this item
CIP75 peptide is used for blocking the activity of CIP75 antibody.