Human erbB-2 peptide (19 aa near C-terminus)
PRSI5153P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:19 aa near C-terminus
- Protein/peptide name:erbB-2 Peptide (19 aa near C-terminus)
- Tested applications:Blocking
erbB-2 peptide is used for blocking the activity of RUNX1 antibody.