Human LRFN1 peptide (18 aa near C-terminus)
PRSI5065P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near C-terminus
- Protein/peptide name:LRFN1 Peptide (18 aa near C-terminus)
- Tested applications:Blocking
LRFN1 peptide is used for blocking the activity of LRFN1 antibody.