Human OCC-1 peptide (16 aa near N-terminus)
Catalog # PRSI4997P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near N-terminus
- Protein/peptide name:OCC-1 Peptide (16 aa near N-terminus)
- Tested applications:Blocking
Specifications
About this item
OCC-1 peptide is used for blocking the activity of OCC-1 antibody.