Human MettL7B peptide (16 aa near C-terminus)
PRSI4881P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near C-terminus
- Protein/peptide name:MettL7B Peptide (16 aa near C-terminus)
- Tested applications:Blocking
MettL7B peptide is used for blocking the activity of MettL7B antibody.