Human NUP155 peptide (18 aa near C-terminus)
Catalog # PRSI4703P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near C-terminus
- Protein/peptide name:NUP155 Peptide (18 aa near C-terminus)
- Tested applications:Blocking
Specifications
About this item
NUP155 peptide is used for blocking the activity of NUP155 antibody.