Order Entry
Export
ContactUsLinkComponent
Human Slitrk1 peptide (18 aa in the center)
Human Slitrk1 peptide (18 aa in the center)
Catalog # PRSI4455P
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Slitrk1 peptide (18 aa in the center)
Catalog # PRSI4455P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 aa in the center
  • Protein/peptide name:
    Slitrk1 Peptide (18 aa in the center)
  • Tested applications:
    Blocking

Specifications

About this item

Slitrk1 peptide is used for blocking the activity of Slitrk1 antibody.