Order Entry
Export
ContactUsLinkComponent
Rift valley fever virus RVF virus peptide (19 aa in the center)
Rift valley fever virus RVF virus peptide (19 aa in the center)
Catalog # PRSI4519P
Supplier:  ProSci Inc.
CAS Number:  
undefined
Rift valley fever virus RVF virus peptide (19 aa in the center)
Catalog # PRSI4519P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Rift valley fever virus
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    19 aa in the center
  • Protein/peptide name:
    RVF virus Peptide (19 aa in the center)
  • Tested applications:
    Blocking

Specifications

About this item

Rift Valley Fever Virus peptide is used for blocking the activity of RVF virus antibody.