Order Entry
Export
ContactUsLinkComponent
 
Human Grik1 peptide (16 aa in the center)
  PRSI4383P
 :  
undefined
Human Grik1 peptide (16 aa in the center)
  PRSI4383P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    16 aa in the center
  • Protein/peptide name:
    Grik1 Peptide (16 aa in the center)
  • Tested applications:
    Blocking

 

 

Grik1 peptide is used for blocking the activity of Grik1 antibody.