Human PTEN peptide (16 aa near C-terminus)
PRSI3515P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near C-terminus
- Protein/peptide name:PTEN Peptide (16 aa near C-terminus)
- Tested applications:Blocking
PTEN peptide is used for blocking the activity of the PTEN antibody.