Human STEAP1 peptide (18 aa near C-terminus)
PRSI4303P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near C-terminus
- Protein/peptide name:STEAP1 Peptide (18 aa near C-terminus)
- Tested applications:Blocking
STEAP1 peptide is used for blocking the activity of STEAP1 antibody.