Order Entry
Export
ContactUsLinkComponent
 
Human IL-33 peptide (19 aa in the center)
  PRSI4273P
 :  
undefined
Human IL-33 peptide (19 aa in the center)
  PRSI4273P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    19 aa in the center
  • Protein/peptide name:
    IL-33 Peptide (19 aa in the center)
  • Tested applications:
    Blocking

 

 

IL-33 peptide is used for blocking the activity of IL-33 antibody.